6-Bromo-3,4-dichloroquinoline

Lieferant: Apollo Scientific
Gefahr

OR309336-1G
APOSOR309336-1GEA 359 EUR
APOSOR309336-1G
6-Bromo-3,4-dichloroquinoline
6-Bromo-3,4-dichloroquinoline
Formel: C₉H₄BrCl₂N
Molecular Weight: 276,945 g/mol
Schmelzpunkt: 128…129 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08437001
CAS-Nummer: 927801-17-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR