4-Amino-3-bromo-8-methoxyquinoline

Lieferant: Apollo Scientific

OR309339-1G
APOSOR309339-1GEA 509 EUR
APOSOR309339-1G
4-Amino-3-bromo-8-methoxyquinoline
4-Amino-3-bromo-8-methoxyquinoline
Formel: C₁₀H₉BrN₂O
Molecular Weight: 253,095 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13619788
CAS-Nummer: 1210642-96-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR