Ethyl-4-chlorchinolin-3-carboxylat

Lieferant: Apollo Scientific

Synonyme: 4-Chloroquinoline-3-carboxylic acid ethyl ester, Ethyl 4-chloroquinoline-3-carboxylate

OR309348-5G
APOSOR309348-5GEA 79.9 EUR
APOSOR309348-5G
Ethyl-4-chlorchinolin-3-carboxylat
Ethyl-4-chlorchinolin-3-carboxylat
Formel: C₁₂H₁₀ClNO₂
Molecular Weight: 235.67 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173407
CAS-Nummer: 13720-94-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR