Ethyl-6-bromo-4-chloroquinoline-3-carboxylate

Lieferant: Apollo Scientific

Synonyme: 6-Bromo-4-chloroquinoline-3-carboxylic acid ethyl ester

OR309351-5G
APOSOR309351-5GEA 162 EUR
APOSOR309351-5G
Ethyl-6-bromo-4-chloroquinoline-3-carboxylate
Ethyl-6-bromo-4-chloroquinoline-3-carboxylate
Formel: C₁₂H₉BrClNO₂
Molecular Weight: 314.563 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173367
CAS-Nummer: 206257-39-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR