Ethyl-8-bromo-4-chloroquinoline-3-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 8-bromo-4-chloroquinoline-3-carboxylate, 8-Bromo-4-chloroquinoline-3-carboxylic acid ethyl ester

OR309352-5G
APOSOR309352-5GEA 190 EUR
APOSOR309352-5G
Ethyl-8-bromo-4-chloroquinoline-3-carboxylate
Ethyl-8-bromo-4-chloroquinoline-3-carboxylate
Formel: C₁₂H₉BrClNO₂
Molecular Weight: 314.563 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173385
CAS-Nummer: 206258-97-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR