4-Amino-6-methoxyquinoline-3-carboxylic acid ethyl ester

Lieferant: Apollo Scientific

OR309353-1G
APOSOR309353-1GEA 359 EUR
APOSOR309353-1G
4-Amino-6-methoxyquinoline-3-carboxylic acid ethyl ester
4-Amino-6-methoxyquinoline-3-carboxylic acid ethyl ester
Formel: C₁₃H₁₄N₂O₃
Molecular Weight: 246,262 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09761113
CAS-Nummer: 908350-29-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR