4-Amino-8-bromoquinoline-3-carboxylic acid ethyl ester

Lieferant: Apollo Scientific

OR309354-1G
APOSOR309354-1GEA 590 EUR
APOSOR309354-1G
4-Amino-8-bromoquinoline-3-carboxylic acid ethyl ester
4-Amino-8-bromoquinoline-3-carboxylic acid ethyl ester
Formel: C₁₂H₁₁BrN₂O₂
Molecular Weight: 295,132 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16987435
CAS-Nummer: 1242260-89-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR