4-Amino-8-bromo-6-methylquinoline-3-carboxylic acid ethyl ester

Lieferant: Apollo Scientific

OR309355-1G
APOSOR309355-1GEA 525 EUR
APOSOR309355-1G
4-Amino-8-bromo-6-methylquinoline-3-carboxylic acid ethyl ester
4-Amino-8-bromo-6-methylquinoline-3-carboxylic acid ethyl ester
Formel: C₁₃H₁₃BrN₂O₂
Molecular Weight: 309.159 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16987452
CAS-Nummer: 1242260-34-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR