4-Amino-6-bromoquinoline-3-carboxylic acid

Lieferant: Apollo Scientific

OR309356-1G
APOSOR309356-1GEA 565 EUR
APOSOR309356-1G
4-Amino-6-bromoquinoline-3-carboxylic acid
4-Amino-6-bromoquinoline-3-carboxylic acid
Formel: C₁₀H₇BrN₂O₂
Molecular Weight: 267,079 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13704745
CAS-Nummer: 1216263-67-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR