trans-2-Amino-1-cyclohexanecarboxylic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR309359-500MG
APOSOR309359-500MGEA
255
EUR
APOSOR309359-500MG
trans-2-Amino-1-cyclohexanecarboxylic acid 97%
trans-2-Amino-1-cyclohexanecarboxylic acid
Formel:
C₇H₁₃NO₂ Molecular Weight: 143,186 g/mol Schmelzpunkt: 255 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00145418 CAS-Nummer: 5691-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...