8-Bromo-6-methylquinoline-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR309361-1G
APOSOR309361-1GEA 499 EUR
APOSOR309361-1G
8-Bromo-6-methylquinoline-3-carboxylic acid
8-Bromo-6-methylquinoline-3-carboxylic acid
Formel: C₁₁H₈BrNO₂
Molecular Weight: 266,091 g/mol
Schmelzpunkt: 242…243 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD18909557
CAS-Nummer: 1296950-68-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR