8-Ethylquinoline-3-carboxamide

Lieferant: Apollo Scientific

OR309362-1G
APOSOR309362-1GEA 498 EUR
APOSOR309362-1G
8-Ethylquinoline-3-carboxamide
8-Ethylquinoline-3-carboxamide
Formel: C₁₂H₁₂N₂O
Molecular Weight: 200,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18909563
CAS-Nummer: 71083-37-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR