3-Amino-7-methylquinoline Dihydrochloride

Lieferant: Apollo Scientific

OR309363-1G
APOSOR309363-1GEA 656 EUR
APOSOR309363-1G
3-Amino-7-methylquinoline Dihydrochloride
3-Amino-7-methylquinoline Dihydrochloride
Formel: C₁₀H₁₀N₂·2ClH
Molecular Weight: 231,122 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18909595
CAS-Nummer: 1296950-60-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR