3-Amino-5,7-dichloroquinoline Dihydrochloride

Lieferant: Apollo Scientific

OR309364-1G
APOSOR309364-1GEA 656 EUR
APOSOR309364-1G
3-Amino-5,7-dichloroquinoline Dihydrochloride
3-Amino-5,7-dichloroquinoline Dihydrochloride
Formel: C₉H₆Cl₂N₂·2ClH
Molecular Weight: 285,985 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18909608
CAS-Nummer: 1296950-84-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR