3-Amino-7-chloro-2-methylquinoline Dihydrochloride

Lieferant: Apollo Scientific

OR309365-1G
APOSOR309365-1GEA 656 EUR
APOSOR309365-1G
3-Amino-7-chloro-2-methylquinoline Dihydrochloride
3-Amino-7-chloro-2-methylquinoline Dihydrochloride
Formel: C₁₀H₉ClN₂·2ClH
Molecular Weight: 265,567 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18909619
CAS-Nummer: 1296950-72-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR