2-Methylquinolin-3-amine

Lieferant: Apollo Scientific

Synonyme: 3-Amino-2-methylquinoline dihydrochloride

OR309366-1G
APOSOR309366-1GEA 389 EUR
APOSOR309366-1G
2-Methylquinolin-3-amine
2-Methylquinolin-3-amine
Formel: C₁₀H₁₀N₂
Molecular Weight: 158.1998 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08705651
CAS-Nummer: 21352-22-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR