8-Bromo-2-methylquinoline-3-carboxylic acid ethyl ester

Lieferant: Apollo Scientific

OR309367-1G
APOSOR309367-1GEA 0 EUR
APOSOR309367-1G
8-Bromo-2-methylquinoline-3-carboxylic acid ethyl ester
8-Bromo-2-methylquinoline-3-carboxylic acid ethyl ester
Formel: C₁₃H₁₂BrNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD20036315
CAS-Nummer: 1333255-21-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR