8-Bromo-2-methylquinoline-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR309368-1G
APOSOR309368-1GEA 533 EUR
APOSOR309368-1G
8-Bromo-2-methylquinoline-3-carboxylic acid
8-Bromo-2-methylquinoline-3-carboxylic acid
Formel: C₁₁H₈BrNO₂
Molecular Weight: 266,091 g/mol
Schmelzpunkt: 201…202 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD20036316
CAS-Nummer: 1333251-88-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR