6-Bromo-3-chloro-4-hydroxy-2-methylquinoline

Lieferant: Apollo Scientific

OR309369-1G
APOSOR309369-1GEA 433 EUR
APOSOR309369-1G
6-Bromo-3-chloro-4-hydroxy-2-methylquinoline
6-Bromo-3-chloro-4-hydroxy-2-methylquinoline
Formel: C₁₀H₇BrClNO
Molecular Weight: 272,526 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD20036381
CAS-Nummer: 1333254-43-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR