4-Bromo-3-chloro-2-methylquinoline

Lieferant: Apollo Scientific

OR309370-1G
APOSOR309370-1GEA 484 EUR
APOSOR309370-1G
4-Bromo-3-chloro-2-methylquinoline
4-Bromo-3-chloro-2-methylquinoline
Formel: C₁₀H₇BrClN
Molecular Weight: 256,526 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20036457
CAS-Nummer: 1333250-12-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR