4-Bromo-3,5,7-trichloro-2-methylquinoline
Lieferant: Apollo Scientific
OR309371-1G
APOSOR309371-1GEA
525
EUR
APOSOR309371-1G
4-Bromo-3,5,7-trichloro-2-methylquinoline
4-Bromo-3,5,7-trichloro-2-methylquinoline
Formel:
C₁₀H₅BrCl₃N Molecular Weight: 325,416 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20036489 CAS-Nummer: 1333255-86-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...