3-Bromo-2,8-dimethyl-4-hydroxyquinoline

Lieferant: Apollo Scientific

OR309372-1G
APOSOR309372-1GEA 383 EUR
APOSOR309372-1G
3-Bromo-2,8-dimethyl-4-hydroxyquinoline
3-Bromo-2,8-dimethyl-4-hydroxyquinoline
Formel: C₁₁H₁₀BrNO
Molecular Weight: 252,107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18157848
CAS-Nummer: 909208-88-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR