3-Bromo-4-chloro-2-methylquinoline
Lieferant: Apollo Scientific
OR309373-1G
APOSOR309373-1GEA
484
EUR
APOSOR309373-1G
3-Bromo-4-chloro-2-methylquinoline
3-Bromo-4-chloro-2-methylquinoline
Formel:
C₁₀H₇BrClN Molecular Weight: 256,526 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13689067 CAS-Nummer: 1333256-98-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...