6-Methoxy-4-(piperazin-1-yl)quinoline Hydrochloride

Lieferant: Apollo Scientific

OR309374-1G
APOSOR309374-1GEA 335 EUR
APOSOR309374-1G
6-Methoxy-4-(piperazin-1-yl)quinoline Hydrochloride
6-Methoxy-4-(piperazin-1-yl)quinoline Hydrochloride
Formel: C₁₄H₁₇N₃O·ClH
Molecular Weight: 279,765 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20036727
CAS-Nummer: 1333249-14-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR