6-Chloro-4-(piperazin-1-yl)quinoline Hydrochloride

Lieferant: Apollo Scientific

OR309375-1G
APOSOR309375-1GEA 335 EUR
APOSOR309375-1G
6-Chloro-4-(piperazin-1-yl)quinoline Hydrochloride
6-Chloro-4-(piperazin-1-yl)quinoline Hydrochloride
Formel: C₁₃H₁₄ClN₃·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD20036733
CAS-Nummer: 1333249-31-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR