6-Methoxy-2-methyl-4-(piperazin-1-yl)quinoline Hydrochloride
Lieferant: Apollo Scientific
OR309376-1G
APOSOR309376-1GEA
335
EUR
APOSOR309376-1G
6-Methoxy-2-methyl-4-(piperazin-1-yl)quinoline Hydrochloride
6-Methoxy-2-methyl-4-(piperazin-1-yl)quinoline Hydrochloride
Formel:
C₁₅H₁₉N₃O·ClH Molecular Weight: 293,792 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20036768 CAS-Nummer: 1333256-88-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...