8-Bromo-2-methyl-4-(piperazin-1-yl)quinoline Hydrochloride

Lieferant: Apollo Scientific

OR309378-1G
APOSOR309378-1GEA 335 EUR
APOSOR309378-1G
8-Bromo-2-methyl-4-(piperazin-1-yl)quinoline Hydrochloride
8-Bromo-2-methyl-4-(piperazin-1-yl)quinoline Hydrochloride
Formel: C₁₄H₁₆BrN₃·ClH
Molecular Weight: 342,662 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20036780
CAS-Nummer: 1333253-23-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR