2-(Piperazin-1-yl)quinolin-3-carbonitrile Hydrochloride

Lieferant: Apollo Scientific

OR309379-1G
APOSOR309379-1GEA 335 EUR
APOSOR309379-1G
2-(Piperazin-1-yl)quinolin-3-carbonitrile Hydrochloride
2-(Piperazin-1-yl)quinolin-3-carbonitrile Hydrochloride
Formel: C₁₄H₁₄N₄·ClH
Molecular Weight: 274,749 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20037065
CAS-Nummer: 1333258-10-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR