6-Methoxy-2-(piperazin-1-yl)quinolin-3-carbonitrile Hydrochloride

Lieferant: Apollo Scientific

OR309380-1G
APOSOR309380-1GEA 335 EUR
APOSOR309380-1G
6-Methoxy-2-(piperazin-1-yl)quinolin-3-carbonitrile Hydrochloride
6-Methoxy-2-(piperazin-1-yl)quinolin-3-carbonitrile Hydrochloride
Formel: C₁₅H₁₆N₄O·ClH
Molecular Weight: 304,775 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20037072
CAS-Nummer: 1333254-61-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR