6-Brom-2-chlor-4-methylchinolin

Lieferant: Apollo Scientific
Achtung

OR309381-250MG
APOSOR309381-250MGEA 72.5 EUR
APOSOR309381-250MG
6-Brom-2-chlor-4-methylchinolin
6-Bromo-2-chloro-4-methylquinoline
Formel: C₁₀H₇BrClN
Molecular Weight: 256,5300 g/mol
Schmelzpunkt: 138…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06254262
CAS-Nummer: 3913-19-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR