2-Chlor-7-methoxy-4-methylchinolin

Lieferant: Apollo Scientific

Synonyme: 7-Methoxy-2-chloro-4-methylquinoline

OR309383-1G
APOSOR309383-1GEA 267 EUR
APOSOR309383-1G
2-Chlor-7-methoxy-4-methylchinolin
2-Chloro-7-methoxy-4-methylquinoline
Formel: C₁₁H₁₀ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD00711757
CAS-Nummer: 97892-67-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR