2,6-Dichloro-4-methylquinoline

Lieferant: Apollo Scientific

OR309384-1G
APOSOR309384-1GEA 351 EUR
APOSOR309384-1G
2,6-Dichloro-4-methylquinoline
2,6-Dichloro-4-methylquinoline
Formel: C₁₀H₇Cl₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD01316208
CAS-Nummer: 90723-71-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR