2-Chloro-6-(2-hydroxyethyl)quinoline-3-carbonitrile
Lieferant: Apollo Scientific
OR309386-1G
APOSOR309386-1GEA
383
EUR
APOSOR309386-1G
2-Chloro-6-(2-hydroxyethyl)quinoline-3-carbonitrile
2-Chloro-6-(2-hydroxyethyl)quinoline-3-carbonitrile
Formel:
C₁₂H₉ClN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD21647758 CAS-Nummer: 1367706-48-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...