2-Hydroxy-4,8-dimethyl-7-bromo-quinoline

Lieferant: Apollo Scientific
Achtung

OR309387-1G
APOSOR309387-1GEA 351 EUR
APOSOR309387-1G
2-Hydroxy-4,8-dimethyl-7-bromo-quinoline
2-Hydroxy-4,8-dimethyl-7-bromo-quinoline
Formel: C₁₁H₁₀BrNO
Molecular Weight: 252,107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21647766
CAS-Nummer: 1367706-85-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR