2-Bromo-4,6-dimethylquinoline

Lieferant: Apollo Scientific

OR309388-1G
APOSOR309388-1GEA 707 EUR
APOSOR309388-1G
2-Bromo-4,6-dimethylquinoline
2-Bromo-4,6-dimethylquinoline
Formel: C₁₁H₁₀BrN
Molecular Weight: 236,108 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21647772
CAS-Nummer: 103858-47-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR