2-Bromo-7-methoxy-4-methylquinoline

Lieferant: Apollo Scientific

OR309389-1G
APOSOR309389-1GEA 0 EUR
APOSOR309389-1G
2-Bromo-7-methoxy-4-methylquinoline
2-Bromo-7-methoxy-4-methylquinoline
Formel: C₁₁H₁₀BrNO
Lagertemperatur: Raumtemperatur
MDL: MFCD21647777
CAS-Nummer: 115486-12-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR