2-Bromo-6-chloro-4-methylquinoline
Lieferant: Apollo Scientific
|
Achtung
|
OR309390-1G
APOSOR309390-1GEA
517
EUR
APOSOR309390-1G
2-Bromo-6-chloro-4-methylquinoline
2-Bromo-6-chloro-4-methylquinoline
Formel:
C₁₀H₇BrClN Molecular Weight: 256,526 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21647781 CAS-Nummer: 18004-64-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...