6-Methoxy-4-methylquinoline hydrochloride

Lieferant: Apollo Scientific

OR309391-1G
APOSOR309391-1GEA 359 EUR
APOSOR309391-1G
6-Methoxy-4-methylquinoline hydrochloride
6-Methoxy-4-methylquinoline hydrochloride
Formel: C₁₁H₁₁NO·ClH
Molecular Weight: 209,672 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00035252
CAS-Nummer: 7461-59-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR