Ethyl 4-chloro-6-methyl-2-quinazolinecarboxylate

Lieferant: Apollo Scientific

OR309394-250MG
APOSOR309394-250MGEA 185 EUR
APOSOR309394-250MG
Ethyl 4-chloro-6-methyl-2-quinazolinecarboxylate
Ethyl 4-chloro-6-methyl-2-quinazolinecarboxylate
Formel: C₁₂H₁₁ClN₂O₂
Molecular Weight: 250,681 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12026444
CAS-Nummer: 620957-95-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR