Ethyl 6-bromo-4-chloroquinazoline-2-carboxylate

Lieferant: Apollo Scientific
Achtung

OR309396-100MG
APOSOR309396-100MGEA 21.9 EUR
APOSOR309396-100MG
Ethyl 6-bromo-4-chloroquinazoline-2-carboxylate
Ethyl 6-bromo-4-chloroquinazoline-2-carboxylate
Formel: C₁₁H₈BrClN₂O₂
Molecular Weight: 315.55 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12026446
CAS-Nummer: 1159976-38-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR