Ethyl 4-chloro-6-methoxyquinazoline-2-carboxylate
Lieferant: Apollo Scientific
OR309398-1G
APOSOR309398-1GEA
0
EUR
APOSOR309398-1G
Ethyl 4-chloro-6-methoxyquinazoline-2-carboxylate
Ethyl 4-chloro-6-methoxyquinazoline-2-carboxylate
Formel:
C₁₂H₁₁ClN₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD10697896 CAS-Nummer: 364385-74-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...