Ethyl 4,7-dichloroquinazoline-2-carboxylate

Lieferant: Apollo Scientific

OR309399-1G
APOSOR309399-1GEA 325 EUR
APOSOR309399-1G
Ethyl 4,7-dichloroquinazoline-2-carboxylate
Ethyl 4,7-dichloroquinazoline-2-carboxylate
Formel: C₁₁H₈Cl₂N₂O₂
Molecular Weight: 271,099 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12675150
CAS-Nummer: 1189106-09-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR