Ethyl 4-chloro-6,7-dimethoxyquinazoline-2-carboxylate
Lieferant: Apollo Scientific
OR309400-1G
APOSOR309400-1GEA
459
EUR
APOSOR309400-1G
Ethyl 4-chloro-6,7-dimethoxyquinazoline-2-carboxylate
Ethyl 4-chloro-6,7-dimethoxyquinazoline-2-carboxylate
Formel:
C₁₃H₁₃ClN₂O₄ Molecular Weight: 296,706 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12675151 CAS-Nummer: 1189105-82-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...