Ethyl 4-chloro-7-methoxyquinazoline-2-carboxylate

Lieferant: Apollo Scientific

OR309401-1G
APOSOR309401-1GEA 359 EUR
APOSOR309401-1G
Ethyl 4-chloro-7-methoxyquinazoline-2-carboxylate
Ethyl 4-chloro-7-methoxyquinazoline-2-carboxylate
Formel: C₁₂H₁₁ClN₂O₃
Molecular Weight: 266,68 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12675152
CAS-Nummer: 1189107-22-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR