Ethyl 7-bromo-4-chloroquinazoline-2-carboxylate

Lieferant: Apollo Scientific

OR309402-1G
APOSOR309402-1GEA 441 EUR
APOSOR309402-1G
Ethyl 7-bromo-4-chloroquinazoline-2-carboxylate
Ethyl 7-bromo-4-chloroquinazoline-2-carboxylate
Formel: C₁₁H₈BrClN₂O₂
Molecular Weight: 315.55 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12675153
CAS-Nummer: 1189105-79-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR