4-Chloro-2-(chloromethyl)quinazoline

Lieferant: Apollo Scientific

OR309405-1G
APOSOR309405-1GEA 484 EUR
APOSOR309405-1G
4-Chloro-2-(chloromethyl)quinazoline
4-Chloro-2-(chloromethyl)quinazoline
Formel: C₉H₆Cl₂N₂
Molecular Weight: 213,063 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07758775
CAS-Nummer: 34637-41-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR