7-Brom-4-chlorchinazolin

Lieferant: Apollo Scientific

OR309406-1G
APOSOR309406-1GEA 0 EUR
APOSOR309406-1G
7-Brom-4-chlorchinazolin
7-Bromo-4-chloroquinazoline
Formel: C₈H₄BrClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06657216
CAS-Nummer: 573675-55-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR