2-(5-Thioxo-1H-1,2,4-triazol-4(5H)-yl)acetic acid

Lieferant: Apollo Scientific

OR309407-1G
APOSOR309407-1GEA 475 EUR
APOSOR309407-1G
2-(5-Thioxo-1H-1,2,4-triazol-4(5H)-yl)acetic acid
2-(5-Thioxo-1H-1,2,4-triazol-4(5H)-yl)acetic acid
Formel: C₄H₅N₃O₂S
Molecular Weight: 159,166 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08437534
CAS-Nummer: 61336-27-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR