(3-Phenyl-5-thioxo-1,5-dihydro-[1,2,4]triazol-4-yl)-acetic acid

Lieferant: Apollo Scientific

OR309409-1G
APOSOR309409-1GEA 475 EUR
APOSOR309409-1G
(3-Phenyl-5-thioxo-1,5-dihydro-[1,2,4]triazol-4-yl)-acetic acid
(3-Phenyl-5-thioxo-1,5-dihydro-[1,2,4]triazol-4-yl)-acetic acid
Formel: C₁₀H₉N₃O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD05721268
CAS-Nummer: 185034-18-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR