3-Amino-2-brom-4-methylpyridin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-2-bromo-4-picoline

OR30941-1G
APOSOR30941-1GEA 21.9 EUR
APOSOR30941-1G
3-Amino-2-brom-4-methylpyridin 97%
3-Amino-2-bromo-4-methylpyridine
Formel: C₆H₇BrN₂
Molecular Weight: 187.04 g/mol
Schmelzpunkt: 78…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04112497
CAS-Nummer: 126325-50-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR